missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (R)-(-)-3-hydroxybutyrate

For synthesis of optically active products

Manufacturer:  Alfa Aesar™ J6340003

 View more versions of this product

Catalog No. AAJ6340003



Specifications

Specifications

Ethyl (R)-(-)-3-hydroxybutyrate
1g
C6H12O3
butanoic acid, 3-hydroxy-, ethyl ester, 3r, ethyl 3r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, ethyl r-3-hydroxybutanoate, ethyl r-3-hydroxybutyrate, r---3-hydroxy-n-butyric acid ethyl ester, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, r-ethyl 3-hydroxybutanoate
CCOC(=O)CC(C)O
132.159
CHEBI:28707
Liquid
Colorless
24915-95-5
MFCD00075386
OMSUIQOIVADKIM-RXMQYKEDSA-N
ethyl (3R)-3-hydroxybutanoate
440030
132.16

Chemical Identifiers

24915-95-5
132.159
OMSUIQOIVADKIM-RXMQYKEDSA-N
440030
ethyl (3R)-3-hydroxybutanoate
C6H12O3
MFCD00075386
butanoic acid, 3-hydroxy-, ethyl ester, 3r, ethyl 3r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, ethyl r-3-hydroxybutanoate, ethyl r-3-hydroxybutyrate, r---3-hydroxy-n-butyric acid ethyl ester, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, r-ethyl 3-hydroxybutanoate
CHEBI:28707
CCOC(=O)CC(C)O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents