Please login to your online account to display your discounted pricing

Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338870050

 View more versions of this product

Catalog No. AC338870050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 208.26
Quantity 5g
Color Clear colorless
Specific Rotation Condition − 10.00 (20.00°C c=neat)
Density 1.0750g/mL
Specific Rotation -9° to -11° (20°C, 589nm) (neat)
Refractive Index 1.5030 to 1.5050 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 90315-82-5
Assay Percent Range 98%
Linear Formula C6H5(CH2)2CH(OH)COOCH2CH3
MDL Number MFCD00077794
Name Note 98%
Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate
Molecular Formula C12H16O3

Specifications

Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate
Name Note 98%
CAS 90315-82-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H16O3
Linear Formula C6H5(CH2)2CH(OH)COOCH2CH3
Formula Weight 208.26
MDL Number MFCD00077794