missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%, ACROS Organics™

Chemical Identifiers

CAS 90315-82-5
Assay Percent Range 98%
Molecular Formula C12H16O3
Linear Formula C6H5(CH2)2CH(OH)COOCH2CH3
Formula Weight 208.26
MDL Number MFCD00077794
Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC338870050 SDS Thermo Fisher Scientific
338870050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90315-82-5
C12H16O3
208.26
Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%
98%
C6H5(CH2)2CH(OH)COOCH2CH3
MFCD00077794
≥97.5%

Specifications

Liquid
5g
− 10.00 (20.00°C c=neat)
-9° to -11° (20°C, 589nm) (neat)
Authentic
90315-82-5
C6H5(CH2)2CH(OH)COOCH2CH3
C12H16O3
≥97.5%
208.26
Clear colorless
1.0750g/mL
1.5030 to 1.5050 (20°C, 589nm)
Glass bottle
98%
MFCD00077794
Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%