Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate
CAS 90315-82-5
Formula Weight 208.25
MDL Number MFCD00077794
Beilstein 3590943
Synonym (R)-2-hydroxy-4-phenylbutyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6150706

 
alfa aesar™
H6150706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6150714

 
alfa aesar™
H6150714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.25
Chemical Name or Material Ethyl (R)-2-hydroxy-4-phenylbutyrate
Refractive Index 1.5040
Physical Form Liquid
Assay 98%
MDL Number MFCD00077794
CAS 90315-82-5
Beilstein 3590943
Synonym (R)-2-hydroxy-4-phenylbutyric acid ethyl ester
Density 1.075g/mL
Boiling Point 212°C
Color Colorless
Quantity 5g