missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (R)-2-hydroxy-4-phenylbutyrate, 98%

Manufacturer:  Alfa Aesar™ H6150706

 View more versions of this product

Catalog No. AAH6150706



Specifications

Specifications

Ethyl (R)-2-hydroxy-4-phenylbutyrate
150°C (302°F)
212°C
5g
C12H16O3
3590943
ZJYKSSGYDPNKQS-LLVKDONJSA-N
ethyl (2R)-2-hydroxy-4-phenylbutanoate
2733848
Liquid
1.075 g/mL
1.504
Colorless
90315-82-5
MFCD00077794
ethyl 2r-2-hydroxy-4-phenylbutanoate, ethyl r---2-hydroxy-4-phenylbutyrate, ethyl r-2-hydroxy-4-phenylbutyrate, qp7iwh2qox, r---2-hydroxy-4-phenylbutyrate ethyl ester, r-2-hydroxy-4-phenyl butyric acid ethyl ester, r-2-hydroxy-4-phenylbutyric acid ethyl ester, r-ethyl 2-hydroxy-4-phenylbutanoate, r-hpbe, unii-qp7iwh2qox
CCOC(=O)C(CCC1=CC=CC=C1)O
208.257
208.25
98%

Chemical Identifiers

90315-82-5
208.257
ZJYKSSGYDPNKQS-LLVKDONJSA-N
2733848
CCOC(=O)C(CCC1=CC=CC=C1)O
C12H16O3
MFCD00077794
ethyl 2r-2-hydroxy-4-phenylbutanoate, ethyl r---2-hydroxy-4-phenylbutyrate, ethyl r-2-hydroxy-4-phenylbutyrate, qp7iwh2qox, r---2-hydroxy-4-phenylbutyrate ethyl ester, r-2-hydroxy-4-phenyl butyric acid ethyl ester, r-2-hydroxy-4-phenylbutyric acid ethyl ester, r-ethyl 2-hydroxy-4-phenylbutanoate, r-hpbe, unii-qp7iwh2qox
ethyl (2R)-2-hydroxy-4-phenylbutanoate
Safety and Handling

Safety and Handling

EINECSNumber : 618-525-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents