missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl pyruvate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1504536

 View more versions of this product

Catalog No. AAA1504536



Description

Description

Solubility
Miscible with water, ethanol and ether.

Notes
Store in a cool place. Incompatible with oxidizing agents.
Specifications

Specifications

Ethyl pyruvate
Pleasant
-58°C
617-35-6
C5H8O3
UN3272
14,8021
XXRCUYVCPSWGCC-UHFFFAOYSA-N
ethyl 2-oxopropanoate
12041
98%
45°C (113°F)
143°C to 145°C
500g
98%
MFCD00009123
1071466
ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
CCOC(=O)C(=O)C
116.116
116.12

Chemical Identifiers

617-35-6
116.116
XXRCUYVCPSWGCC-UHFFFAOYSA-N
12041
CCOC(=O)C(=O)C
C5H8O3
MFCD00009123
ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
ethyl 2-oxopropanoate
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

storageNote1 : Store cold

Recommended Storage : 2° to 8°C

Documents