missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl pyrrolidinoacetate, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 35092-0250

 View more versions of this product

Catalog No. AC350920250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
157.21
Clear light yellow
1.4460 to 1.4480 (20°C, 589nm)
Authentic
C8H15NO2
MFCD00012085
22041-19-6
Liquid
25g
20, V,1, 335
Glass bottle
Ethyl pyrrolidinoacetate
98%
98%

Chemical Identifiers

22041-19-6
C8H15NO2
MFCD00012085
≥97.5%
98%
157.21
Ethyl pyrrolidinoacetate, 98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : UX9670300