missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl pyrrolidinoacetate, 98%, ACROS Organics™

Chemical Identifiers

CAS 22041-19-6
Assay Percent Range 98%
Molecular Formula C8H15NO2
Formula Weight 157.21
MDL Number MFCD00012085
Chemical Name or Material Ethyl pyrrolidinoacetate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC350920050 SDS Thermo Fisher Scientific
35092-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350920250 SDS Fisher Scientific Acros
35092-0250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

22041-19-6
C8H15NO2
MFCD00012085
≥97.5%
98%
157.21
Ethyl pyrrolidinoacetate, 98%

Specifications

Clear light yellow
22041-19-6
Glass bottle
Liquid
C8H15NO2
MFCD00012085
Ethyl pyrrolidinoacetate, 98%
5g
Authentic
1.4460 to 1.4480 (20°C, 589nm)
98%
157.21
20, V,1, 335
≥97.5%

Safety and Handling

RTECSNumber : UX9670300