missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl propionylacetate, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC232910010

 View more versions of this product

Catalog No. AC232910010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl propionylacetate
77°C
Glass bottle
1.01
Colorless to Yellow
4949-44-4
C7H12O3
MFCD00009317
UDRCONFHWYGWFI-UHFFFAOYSA-N
ethyl 3-oxopentanoate
78656
Liquid
1.0100g/mL
Authentic
1.4220 to 1.4240
83.0°C to 84.0°C (12.0mmHg)
1mL
98% min. (GC)
C2H5COCH2CO2C2H5
3-oxo-pentanoic acid ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxovaleric acid ethyl ester, ethyl 3-ketopentanoate, ethyl 3-oxo-n-valerate, ethyl 3-oxovalerate, ethyl propionylacetate, ethylpropanylacetate, pentanoic acid, 3-oxo-, ethyl ester, propionylacetic acid ethyl ester
CCC(=O)CC(=O)OCC
144.17
144.17
98+%

Chemical Identifiers

4949-44-4
144.17
UDRCONFHWYGWFI-UHFFFAOYSA-N
78656
CCC(=O)CC(=O)OCC
C7H12O3
MFCD00009317
3-oxo-pentanoic acid ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxovaleric acid ethyl ester, ethyl 3-ketopentanoate, ethyl 3-oxo-n-valerate, ethyl 3-oxovalerate, ethyl propionylacetate, ethylpropanylacetate, pentanoic acid, 3-oxo-, ethyl ester, propionylacetic acid ethyl ester
ethyl 3-oxopentanoate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 225-593-5

SDS Documents