Learn More
Ethyl propionate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 015498A1
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl propionate | |
-73°C | |
99°C | |
C5H10O2 | |
MFCD00009308 | |
UN1195 | |
14,3847 | |
FKRCODPIKNYEAC-UHFFFAOYSA-N | |
ethyl propanoate | |
7749 | |
102.13 |
105-37-3 | |
0.891 g/mL | |
12°C (54°F) | |
1.385 | |
1 kg | |
506287 | |
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester | |
CCC(=O)OCC | |
102.133 | |
CHEBI:41330 | |
Liquid |
Chemical Identifiers
105-37-3 | |
102.133 | |
FKRCODPIKNYEAC-UHFFFAOYSA-N | |
7749 | |
ethyl propanoate |
C5H10O2 | |
MFCD00009308 | |
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester | |
CHEBI:41330 | |
CCC(=O)OCC |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL PROPIONATE
EINECSNumber : 203-291-4
RTECSNumber : UF3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only