missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 410370250

 View more versions of this product

Catalog No. AC410370250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl propenyl ether
−18°C
Glass bottle
0.778
Colorless to Yellow
98%
97.5% min. (GC)
CH3CHCHOCH2CH3
01,385
Solubility in water: soluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
CCOC=CC
86.13
86.13
98%
0.7780g/mL
Authentic
1.3940 to 1.4000
67.0°C to 76.0°C
25g
928-55-2
C5H10O
MFCD00015184
1-ethoxy-1-propene, 1-ethoxypropene, 1-propene, 1-ethoxy, e-1-ethoxyprop-1-ene, ether, ethyl propenyl, ethyl 1-propenyl ether, ethyl propenyl ether, ethyl-1-propenyl ether, propenyl ethyl ether, unii-jh817due12
XDHOEHJVXXTEDV-HWKANZROSA-N
(E)-1-ethoxyprop-1-ene
5365091
Liquid

Chemical Identifiers

928-55-2
86.13
XDHOEHJVXXTEDV-HWKANZROSA-N
5365091
CCOC=CC
C5H10O
MFCD00015184
1-ethoxy-1-propene, 1-ethoxypropene, 1-propene, 1-ethoxy, e-1-ethoxyprop-1-ene, ether, ethyl propenyl, ethyl 1-propenyl ether, ethyl propenyl ether, ethyl-1-propenyl ether, propenyl ethyl ether, unii-jh817due12
(E)-1-ethoxyprop-1-ene
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-176-

SDS Documents