missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Potassium Oxalate 95.0+%, TCI America™
Supplier: TCI America E118425G
Specifications
Ethyl Potassium Oxalate | |
220°C | |
C4H5KO4 | |
25 g | |
RLPQQBNSTHRHEK-UHFFFAOYSA-M | |
potassium;2-ethoxy-2-oxoacetate | |
23678856 | |
≥95.0% (T) |
1906-57-6 | |
White | |
MFCD00045861 | |
ethyl potassium oxalate, potassium ethyl oxalate, ethanedioic acid, monoethyl ester, potassium salt, potassium 2-ethoxy-2-oxoacetate, oxalic acid 1-ethyl 2-potassium salt, ethanedioic acid, 1-ethyl ester, potassium salt 1:1, potassium ethoxy oxo acetate, ambkkkkk567, oxalic acid, monoethyl ester, potassium salt, potassium monoethyl oxalate | |
CCOC(=O)C(=O)[O-].[K+] | |
156.178 | |
156.18 | |
Crystalline Powder |
Chemical Identifiers
1906-57-6 | |
156.178 | |
RLPQQBNSTHRHEK-UHFFFAOYSA-M | |
23678856 | |
CCOC(=O)C(=O)[O-].[K+] |
C4H5KO4 | |
MFCD00045861 | |
ethyl potassium oxalate, potassium ethyl oxalate, ethanedioic acid, monoethyl ester, potassium salt, potassium 2-ethoxy-2-oxoacetate, oxalic acid 1-ethyl 2-potassium salt, ethanedioic acid, 1-ethyl ester, potassium salt 1:1, potassium ethoxy oxo acetate, ambkkkkk567, oxalic acid, monoethyl ester, potassium salt, potassium monoethyl oxalate | |
potassium;2-ethoxy-2-oxoacetate |
Safety and Handling
TSCA : Yes