Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl piperidine-1-acetate, 98%

$37.68 - $471.62

Chemical Identifiers

Chemical Name or Material Ethyl piperidine-1-acetate
CAS 23853-10-3
Molecular Formula C9H17NO2
Formula Weight 171.24
MDL Number MFCD00134496
Beilstein 123142
Synonym Piperidine-1-acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1366306

  SDS
alfa aesar™
A1366306
5g Each for $37.68

AAA1366322

  SDS
alfa aesar™
A1366322
100g Each for $471.62
Description & Specifications

Specifications

Formula Weight 171.24
Chemical Name or Material Ethyl piperidine-1-acetate
Assay 98%
Molecular Formula C9H17NO2
MDL Number MFCD00134496
CAS 23853-10-3
Beilstein 123142
Synonym Piperidine-1-acetic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 91°C (195°F)
Boiling Point 109° to 111°C (25mmHg)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.