missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl piperazine-1-carboxylate, 99%

$44.01 - $131.92

Chemical Identifiers

CAS 120-43-4
Molecular Formula C7H14N2O2
Formula Weight 158.2
MDL Number MFCD00005964
Beilstein 125780
Synonym N-Carboethoxypiperazine; 1-Ethoxycarbonylpiperazine
Chemical Name or Material Ethyl piperazine-1-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1357314 SDS Alfa Aesar™
A1357314
25g Each for $44.01
AAA1357322 SDS Alfa Aesar™
A1357322
100g Each for $131.92
AAA1357336 SDS Alfa Aesar™
A1357336
500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

120-43-4
158.2
125780
Ethyl piperazine-1-carboxylate
C7H14N2O2
MFCD00005964
N-Carboethoxypiperazine; 1-Ethoxycarbonylpiperazine

Specifications

120-43-4
158.2
125780
237°C
128°C (262°F)
99%
C7H14N2O2
MFCD00005964
N-Carboethoxypiperazine; 1-Ethoxycarbonylpiperazine
25g
Ethyl piperazine-1-carboxylate

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TL1378000

TSCA : TSCA