Learn More
Ethyl pipecolinate, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 189580250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl pipecolinate | |
1.0060g/mL | |
46°C | |
98% min. (GC) | |
C8H16NO2 | |
MFCD00005980 | |
22, 7 | |
ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720 | |
SZIKRGHFZTYTIT-ZETCQYMHSA-O | |
ethyl piperidine-2-carboxylate | |
27517 | |
98+% |
15862-72-3 | |
216°C to 217°C | |
Authentic | |
Glass bottle | |
1.4550 to 1.457 | |
25g | |
1.006 | |
Solubility in water: miscible | |
CCOC(=O)[C@@H]1CCCC[NH2+]1 | |
158.22 | |
157.21 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Warning
EINECSNumber : 239-990-6
RUO â Research Use Only