missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl pipecolinate, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC189580050

 View more versions of this product

Catalog No. AC189580050


 

Specifications Description & Specifications

Specifications

Ethyl pipecolinate
5g
Authentic
Glass bottle
Liquid
98+%
157.21
22, 7
Clear colorless to light yellow
15862-72-3
1.0060g/mL
1.4550 to 1.4570 (20°C, 589nm)
≥98.0%
C8H15NO2
MFCD00005980
Ethyl 2-piperidinecarboxylate

Chemical Identifiers

15862-72-3
C8H15NO2
MFCD00005980
Ethyl pipecolinate
98+%
157.21
Ethyl 2-piperidinecarboxylate
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS P Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning