Please login to your online account to display your discounted pricing

Ethyl pipecolinate, 98+%, ACROS Organics™

$91.40 - $91.40

Chemical Identifiers

Chemical Name or Material Ethyl pipecolinate
CAS 15862-72-3
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H15NO2
Formula Weight 157.21
MDL Number MFCD00005980
Synonym Ethyl 2-piperidinecarboxylate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189580050

  SDS
thermo fisher scientific
AC189580050
Glass bottle 5g Each for $91.40

AC189580250

  SDS
thermo fisher scientific
AC189580250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 157.21
Quantity 5g
Color Clear colorless to light yellow
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Density 1.0060g/mL
Beilstein 22, 7
Chemical Name or Material Ethyl pipecolinate
Refractive Index 1.4550 to 1.4570 (20°C, 589nm)
Assay Percent Range 98+%
Molecular Formula C8H15NO2
MDL Number MFCD00005980
CAS 15862-72-3
Synonym Ethyl 2-piperidinecarboxylate
Health Hazard 1 Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.