Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (phenylthio)acetate, 97%

$267.84 - $267.84

Chemical Identifiers

Chemical Name or Material Ethyl (phenylthio)acetate
CAS 7605-25-6
Molecular Formula C10H12O2S
Formula Weight 196.27
MDL Number MFCD00145107
Synonym (Phenylthio)acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1866409

 
alfa aesar™
A1866409
10g Each for $267.84
Description & Specifications

Specifications

Formula Weight 196.27
Chemical Name or Material Ethyl (phenylthio)acetate
Assay 97%
Molecular Formula C10H12O2S
MDL Number MFCD00145107
CAS 7605-25-6
Synonym (Phenylthio)acetic acid ethyl ester
Boiling Point 143° to 145°C (14mmHg)
Quantity 10g