missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl phenylpropiolate, 98+%

Manufacturer:  Alfa Aesar™ L0060314

 View more versions of this product

Catalog No. AAL0060314



Description

Description

Applications
Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

Ethyl phenylpropiolate
>110°C (230°F)
1.552
25g
C11H10O2
639637
Insoluble in water.
CCOC(=O)C#CC1=CC=CC=C1
174.199
174.2
1.055
Odorless
152°C to 153°C (21mmHg)
2216-94-6
MFCD00009185
2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenyl-2-propynoate, ethyl 3-phenylpropiolate, ethyl 3-phenylpropynoate, ethyl phenylacetylenecarboxylate, ethyl phenylpropiolate, ethyl phenylpropriolate, ethyl phenylpropynoate, ethylphenylpropiolate, phenylpropiolic acid ethyl ester
ACJOYTKWHPEIHW-UHFFFAOYSA-N
ethyl 3-phenylprop-2-ynoate
91516
≥98%

Chemical Identifiers

2216-94-6
174.199
ACJOYTKWHPEIHW-UHFFFAOYSA-N
91516
CCOC(=O)C#CC1=CC=CC=C1
C11H10O2
MFCD00009185
2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenyl-2-propynoate, ethyl 3-phenylpropiolate, ethyl 3-phenylpropynoate, ethyl phenylacetylenecarboxylate, ethyl phenylpropiolate, ethyl phenylpropriolate, ethyl phenylpropynoate, ethylphenylpropiolate, phenylpropiolic acid ethyl ester
ethyl 3-phenylprop-2-ynoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 218-703-8

RTECSNumber : UE0110000

TSCA : No

Recommended Storage : Keep cold

Documents