missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Phenyl Sulfone, TCI America™

Manufacturer:  TCI America E042710G

 View more versions of this product

Catalog No. E042710G



Specifications

Specifications

Ethyl Phenyl Sulfone
White-Yellow
10g
C8H10O2S
ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
CCS(=O)(=O)C1=CC=CC=C1
170.226
170.23
≥97.0% (GC)
103°C
42°C
599-70-2
MFCD00025081
VBQUDDWATQWCPP-UHFFFAOYSA-N
ethylsulfonylbenzene
69032
Crystal-Powder at 20°C

Chemical Identifiers

599-70-2
170.226
VBQUDDWATQWCPP-UHFFFAOYSA-N
69032
CCS(=O)(=O)C1=CC=CC=C1
C8H10O2S
MFCD00025081
ethyl phenyl sulfone, ethylsulfonyl benzene, ethylphenyl sulfone, ethyl phenyl sulphone, sulfone, ethyl phenyl, ethylsulphonyl benzene, ethylphenylsulfone, benzene, ethylsulfonyl, ethanesulfonyl benzene, 3blu
ethylsulfonylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents