Please login to your online account to display your discounted pricing

Ethyl phenyl sulfone, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl phenyl sulfone
Name Note 98%
CAS 599-70-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00025081
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351010010

  SDS
thermo fisher scientific
351010010
Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 170.23
Quantity 1g
Color White to light yellow
EINECS Number 209-972-2
Melting Point 39° to 43°C
Chemical Name or Material Ethyl phenyl sulfone
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H10O2S
MDL Number MFCD00025081
CAS 599-70-2
Infrared Spectrum Authentic
Packaging Glass Bottle