Please login to your online account to display your discounted pricing

Ethyl phenyl sulfone, 98%, ACROS Organics™


Quantity 1g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  351010010

 View more versions of this product

Catalog No. AC351010010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 170.23
Quantity 1g
Color White to light yellow
EINECS Number 209-972-2
Melting Point 39° to 43°C
Chemical Name or Material Ethyl phenyl sulfone
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H10O2S
MDL Number MFCD00025081
CAS 599-70-2
Infrared Spectrum Authentic
Packaging Glass Bottle

Specifications

Chemical Name or Material Ethyl phenyl sulfone
Name Note 98%
CAS 599-70-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00025081