Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl phenyl sulfone, 98+%

$76.70 - $852.82

Chemical Identifiers

Chemical Name or Material Ethyl phenyl sulfone
CAS 599-70-2
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00025081
Beilstein 2045095
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1071006

  SDS
alfa aesar™
A1071006
5g Each for $76.70

AAA1071014

  SDS
alfa aesar™
A1071014
25g Each for $254.65

AAA1071022

  SDS
alfa aesar™
A1071022
100g Each for $852.82
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material Ethyl phenyl sulfone
Assay ≥98%
Molecular Formula C8H10O2S
MDL Number MFCD00025081
CAS 599-70-2
Beilstein 2045095
Melting Point 40° to 44°C
Quantity 5g