Please login to your online account to display your discounted pricing

Ethyl phenoxyacetate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334940050

 View more versions of this product

Catalog No. AC334940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 180.2
Quantity 5g
Color Clear yellow
EINECS Number 219-867-3
TSCA TSCA
Density 1.1000g/mL
Refractive Index 1.5045 to 1.5065 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H12O3
Assay Percent Range 98%
Name Note 98%
CAS 2555-49-9
Chemical Name or Material Ethyl phenoxyacetate
MDL Number MFCD00026895

Specifications

Chemical Name or Material Ethyl phenoxyacetate
Name Note 98%
CAS 2555-49-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00026895