Please login to your online account to display your discounted pricing

Ethyl phenoxyacetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl phenoxyacetate
Name Note 98%
CAS 2555-49-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00026895
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334940050

  SDS
thermo fisher scientific
334940050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334940250

  SDS
thermo fisher scientific
AC334940250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC334941000

  SDS
thermo fisher scientific
AC334941000
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 180.2
Quantity 5g
Color Clear yellow
EINECS Number 219-867-3
TSCA TSCA
Density 1.1000g/mL
Refractive Index 1.5045 to 1.5065 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10H12O3
Assay Percent Range 98%
Name Note 98%
CAS 2555-49-9
Chemical Name or Material Ethyl phenoxyacetate
MDL Number MFCD00026895