Please login to your online account to display your discounted pricing

Ethyl phenoxyacetate, 98%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC334941000

 View more versions of this product

Catalog No. AC334941000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.2
Assay ≥97.5%
Physical Form Liquid
Quantity 100g
Color Clear yellow
TSCA TSCA
EINECS Number 219-867-3
Refractive Index 1.5045 to 1.5065 (20°C, 589nm)
Chemical Name or Material Ethyl phenoxyacetate
Assay Percent Range 98%
Infrared Spectrum Authentic
Molecular Formula C10H12O3
CAS 2555-49-9
MDL Number MFCD00026895
Name Note 98%

Specifications

Chemical Name or Material Ethyl phenoxyacetate
Name Note 98%
CAS 2555-49-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00026895