Please login to your online account to display your discounted pricing

Ethyl pentafluoropropionate, 98%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  311840050

 View more versions of this product

Catalog No. AC311840050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 192.09
Quantity 5g
Color Clear colorless
EINECS Number 207-043-6
TSCA TSCA
Density 1.299g/mL
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.3000 to 1.3200 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material Ethyl pentafluoropropionate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C5H5F5O2
Linear Formula CF5COCH5
CAS 426-65-3
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Ethyl pentafluoropropionate
Name Note 98%
CAS 426-65-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H5F5O2
Linear Formula CF5COCH5
Formula Weight 192.09
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents