missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Palmitate, TCI America™

Manufacturer:  TCI America P0003500ML

 View more versions of this product

Catalog No. P0003500ML



Specifications

Specifications

Ethyl Palmitate
500mL
C18H36O2
ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
CCCCCCCCCCCCCCCC(=O)OCC
284.484
CHEBI:84932
Crystal-Lump at 20°C
White-Yellow
628-97-7
MFCD00008996
XIRNKXNNONJFQO-UHFFFAOYSA-N
ethyl hexadecanoate
12366
284.48
≥97.0% (GC)

Chemical Identifiers

628-97-7
284.484
XIRNKXNNONJFQO-UHFFFAOYSA-N
12366
ethyl hexadecanoate
C18H36O2
MFCD00008996
ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
CHEBI:84932
CCCCCCCCCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

Documents