missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl palmitate, 97%

Manufacturer:  Alfa Aesar™ A1569436

 View more versions of this product

Catalog No. AAA1569436



Description

Description

Applications
Ethyl palmitate acts as a lubricant on the skin’s surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.

Solubility
Miscible with ethanol, dimethyl sulfoxide, chloroform and ethyl acetate. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Ethyl palmitate
192°C to 193°C (10mmHg)
500g
C18H36O2
1782663
XIRNKXNNONJFQO-UHFFFAOYSA-N
ethyl hexadecanoate
12366
284.48
178°C (352°F)
22°C to 26°C
628-97-7
MFCD00008996
ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
CCCCCCCCCCCCCCCC(=O)OCC
284.484
CHEBI:84932
97%

Chemical Identifiers

628-97-7
284.484
XIRNKXNNONJFQO-UHFFFAOYSA-N
12366
ethyl hexadecanoate
C18H36O2
MFCD00008996
ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
CHEBI:84932
CCCCCCCCCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents