Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl p-tolylacetate, 97+%

$61.64 - $266.70

Chemical Identifiers

Chemical Name or Material Ethyl p-tolylacetate
CAS 14062-19-2
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00009181
Beilstein 2045927
Synonym p-Tolylacetic acid ethyl ester; Ethyl 4-methylphenylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1671809

 
alfa aesar™
A1671809
10g Each for $61.64

AAA1671818

 
alfa aesar™
A1671818
50g Each for $266.70
Description & Specifications

Specifications

Formula Weight 178.23
Beilstein 2045927
Flash Point 109°C (228°F)
Boiling Point 240° to 241°C
Quantity 10g
CAS 14062-19-2
Assay ≥97%
Synonym p-Tolylacetic acid ethyl ester; Ethyl 4-methylphenylacetate
Molecular Formula C11H14O2
MDL Number MFCD00009181
Chemical Name or Material Ethyl p-tolylacetate