Please login to your online account to display your discounted pricing

Ethyl p-toluenesulfonate 85%, pract., ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  410452500

 View more versions of this product

Catalog No. AC410452500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 200.25
Color Clear colorless to brownish
Quantity 250g
Density 1.1740g/mL
Beilstein 11, 99
Merck Index 15, 3907
Linear Formula CH3C6H4SO3C2H5
Chemical Name or Material Ethyl p-toluenesulfonate
Assay Percent Range 97%
Molecular Formula C9 H12 O3 S
MDL Number MFCD00009100
Synonym 4-Methylbenzenesulfonic acid, ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
CAS 80-40-0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

Specifications

Chemical Name or Material Ethyl p-toluenesulfonate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9 H12 O3 S
Linear Formula CH3C6H4SO3C2H5
Formula Weight 200.25
MDL Number MFCD00009100
Synonym 4-Methylbenzenesulfonic acid, ethyl ester
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents