missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl p-toluate, 98+%, Thermo Scientific Chemicals
$219.88 - $219.88
Chemical Identifiers
CAS | 94-08-6 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD00009117 |
InChI Key | NWPWRAWAUYIELB-UHFFFAOYSA-N |
Synonym | ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate |
PubChem CID | 66743 |
IUPAC Name | ethyl 4-methylbenzoate |
SMILES | CCOC(=O)C1=CC=C(C=C1)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1056830
|
Thermo Scientific Chemicals
A1056830 |
250 g |
Each for $219.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
94-08-6 | |
164.204 | |
NWPWRAWAUYIELB-UHFFFAOYSA-N | |
66743 | |
CCOC(=O)C1=CC=C(C=C1)C |
C10H12O2 | |
MFCD00009117 | |
ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate | |
ethyl 4-methylbenzoate |
Specifications
94-08-6 | |
232°C | |
C10H12O2 | |
MFCD00009117 | |
1863756 | |
NWPWRAWAUYIELB-UHFFFAOYSA-N | |
ethyl 4-methylbenzoate | |
66743 | |
≥98% |
1.025 | |
99°C (210°F) | |
1.508 | |
250 g | |
ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate | |
CCOC(=O)C1=CC=C(C=C1)C | |
164.204 | |
164.21 | |
Ethyl p-toluate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-301-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only