Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl oxanilate, 98%

$32.26 - $563.53

Chemical Identifiers

Chemical Name or Material Ethyl oxanilate
CAS 1457-85-8
Molecular Formula C10H11NO3
Formula Weight 193.2
MDL Number MFCD00009118
Beilstein 2098703
Synonym Oxanilic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2328506

  SDS
alfa aesar™
B2328506
5g Each for $32.26

AAB2328514

  SDS
alfa aesar™
B2328514
25g Each for $183.82

AAB2328522

  SDS
alfa aesar™
B2328522
100g Each for $563.53
Description & Specifications

Specifications

Formula Weight 193.2
Chemical Name or Material Ethyl oxanilate
Assay 98%
Molecular Formula C10H11NO3
MDL Number MFCD00009118
CAS 1457-85-8
Beilstein 2098703
Synonym Oxanilic acid ethyl ester
Melting Point 64° to 70°C
Quantity 5g