missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1465322

 View more versions of this product

Catalog No. AAA1465322



Description

Description

Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxalyl chloride is used in the synthesis of 2-oxo-3-alkenoic esters and alpha-keto ester in good yield. It acts as a reactant in the preparation of oxyoxalamide derivatives as an epoxide hydrolase inhibitor.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and alcohols.
Specifications

Specifications

Ethyl oxalyl chloride
1.226
41°C (105°F)
1.416
MFCD00000706
UN2920
Moisture sensitive
Slightly miscible with water.
CCOC(=O)C(=O)Cl
136.531
136.53
4755-77-5
134°C to 135°C
C4H5ClO3
ClCOCO2CH2CH3
100 g
506725
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
OWZFULPEVHKEKS-UHFFFAOYSA-N
ethyl 2-chloro-2-oxoacetate
20884
98%

Chemical Identifiers

4755-77-5
136.531
OWZFULPEVHKEKS-UHFFFAOYSA-N
20884
CCOC(=O)C(=O)Cl
C4H5ClO3
MFCD00000706
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
ethyl 2-chloro-2-oxoacetate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226-H335
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

EINECSNumber : 225-285-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only