Please login to your online account to display your discounted pricing

Ethyl orange sodium salt, 92%, ACROS Organics™

$45.11 - $45.11

Chemical Identifiers

Chemical Name or Material Ethyl Orange sodium salt
Name Note Pure
CAS 62758-12-7
Purity Grade Pure
Assay Percent Range 92%
Molecular Formula C16H18N3NaO3S
Formula Weight 355.38
MDL Number MFCD00007503
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC410300100

  SDS
thermo fisher scientific
410300100
Glass bottle 10g Each for $45.11
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 355.38
Quantity 10g
Color Orange
Purity Grade Pure
Chemical Name or Material Ethyl Orange sodium salt
Name Note Pure
Assay Percent Range 92%
MDL Number MFCD00007503
CAS 62758-12-7
Infrared Spectrum Authentic
Packaging Glass bottle
UV_Vis 90% min.
Visual Transition Interval from pH 3.5 (red) to pH 4.8 (yellow)
Molecular Formula C16H18N3NaO3S

Indicator