Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl Orange, 98%

$34.49 - $112.51

Chemical Identifiers

Chemical Name or Material Ethyl Orange
CAS 13545-67-0
Molecular Formula C16H19N3O3S
Formula Weight 333.41
MDL Number MFCD00007503
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1757809

 
alfa aesar™
A1757809
10g Each for $34.49

AAA1757818

 
alfa aesar™
A1757818
50g Each for $112.51
Description & Specifications

Specifications

Formula Weight 333.41
Chemical Name or Material Ethyl Orange
Quantity 10g
TSCA TSCA
Assay 98%
Molecular Formula C16H19N3O3S
MDL Number MFCD00007503
CAS 13545-67-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.