missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oleate, tech. 70%

CAS: 111-62-6 | C20H38O2 | 310.522 g/mol

Supplier:  Thermo Scientific Chemicals A156010B

 View more versions of this product

Catalog No. AAA156010B


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl oleate
-32°C
205°C to 208°C
C20H38O2
CH3(CH2)7CH=CH(CH2)7CO2CH2CH3
1000 g
Air Sensitive
LVGKNOAMLMIIKO-QXMHVHEDSA-N
ethyl (Z)-octadec-9-enoate
5363269
310.53
Technical
111-62-6
0.87
>110°C (230°F)
1.451
MFCD00009579
1727318
ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
CCCCCCCCC=CCCCCCCCC(=O)OCC
310.522
CHEBI:84940
70%

Chemical Identifiers

111-62-6
310.522
LVGKNOAMLMIIKO-QXMHVHEDSA-N
5363269
ethyl (Z)-octadec-9-enoate
C20H38O2
MFCD00009579
ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
CHEBI:84940
CCCCCCCCC=CCCCCCCCC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-889-5

RTECSNumber : RG3715000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only