Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl o-tolylacetate, 98%

$88.97 - $1,349.95

Chemical Identifiers

Chemical Name or Material Ethyl o-tolylacetate
CAS 40291-39-2
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00009179
Beilstein 879835
Synonym o-Tolylacetic acid ethyl ester; Ethyl 2-methylphenylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1195509

 
alfa aesar™
A1195509
10g Each for $88.97

AAA1195518

 
alfa aesar™
A1195518
50g Each for $341.72

AAA1195530

 
alfa aesar™
A1195530
250g Each for $1,349.95
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Ethyl o-tolylacetate
Assay 98%
Molecular Formula C11H14O2
MDL Number MFCD00009179
CAS 40291-39-2
Beilstein 879835
Synonym o-Tolylacetic acid ethyl ester; Ethyl 2-methylphenylacetate
Flash Point 106°C (222°F)
Boiling Point 238° to 239°C
Quantity 10g