missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl o-tolylacetate, 98%

Manufacturer:  Alfa Aesar™ A1195518

 View more versions of this product

Catalog No. AAA1195518



Specifications

Specifications

Ethyl o-tolylacetate
238°C to 239°C
40291-39-2
MFCD00009179
ethyl o-tolylacetate, ethyl 2-o-tolyl acetate, ethyl 2-2-methylphenyl acetate, 2-methylphenylacetic acid ethyl ester, o-tolylacetic acid ethyl ester, ethyl o-methylphenylacetate, ethyl 2-methylphenylacetate, benzeneacetic acid, 2-methyl-, ethyl ester, toluene=ethyl acetate, ethyl 2-o-tolylacetate
CCOC(=O)CC1=CC=CC=C1C
178.231
178.23
106°C (222°F)
50g
C11H14O2
879835
HTDQOYWJDUMNHX-UHFFFAOYSA-N
ethyl 2-(2-methylphenyl)acetate
96528
98%

Chemical Identifiers

40291-39-2
178.231
HTDQOYWJDUMNHX-UHFFFAOYSA-N
96528
CCOC(=O)CC1=CC=CC=C1C
C11H14O2
MFCD00009179
ethyl o-tolylacetate, ethyl 2-o-tolyl acetate, ethyl 2-2-methylphenyl acetate, 2-methylphenylacetic acid ethyl ester, o-tolylacetic acid ethyl ester, ethyl o-methylphenylacetate, ethyl 2-methylphenylacetate, benzeneacetic acid, 2-methyl-, ethyl ester, toluene=ethyl acetate, ethyl 2-o-tolylacetate
ethyl 2-(2-methylphenyl)acetate
SDS Documents