missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate, 98+%

Manufacturer:  Alfa Aesar™ H5547003

 View more versions of this product

Catalog No. AAH5547003



Specifications

Specifications

Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate
1g
C13H19NO4S
2144156
KQCBSWBQAXTILK-WYMLVPIESA-N
ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate
6282444
≥98%
54°C to 56°C
38202-27-6
MFCD00009244
ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate
CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C
285.358
285.36

Chemical Identifiers

38202-27-6
285.358
KQCBSWBQAXTILK-WYMLVPIESA-N
6282444
CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C
C13H19NO4S
MFCD00009244
ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate
ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate
Safety and Handling

Safety and Handling

EINECSNumber : 253-825-5

TSCA : No

Recommended Storage : Keep cold

Documents