missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5547014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate | |
54°C to 56°C | |
MFCD00009244 | |
2144156 | |
KQCBSWBQAXTILK-WYMLVPIESA-N | |
ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate | |
6282444 | |
≥98% |
38202-27-6 | |
C13H19NO4S | |
25 g | |
ethyl o-mesitylsulfonylacetohydroxamate, ethyl n-mesitylsulfonyl oxyacetimidate, ethyl o-methylsulphonylacetohydroxamate, ethyl o-2-mesitylenesulfonyl acethydroxamate, o-mesitylsulfonylacetohydroxamic acid ethyl ester, ethyl 1z-n-mesitylsulfonyl oxy ethanimidoate, ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate, kqcbswbqaxtilk-owbhpgmisa-n, z-ethyl n-mesitylsulfonyloxyacetimidate, z-ethyl n-mesitylsulfonyl oxyacetimidate | |
CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C | |
285.358 | |
285.36 |
Chemical Identifiers
38202-27-6 | |
285.358 | |
KQCBSWBQAXTILK-WYMLVPIESA-N | |
6282444 | |
CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C |
C13H19NO4S | |
MFCD00009244 | |
ethyl o-mesitylsulfonylacetohydroxamate, ethyl n-mesitylsulfonyl oxyacetimidate, ethyl o-methylsulphonylacetohydroxamate, ethyl o-2-mesitylenesulfonyl acethydroxamate, o-mesitylsulfonylacetohydroxamic acid ethyl ester, ethyl 1z-n-mesitylsulfonyl oxy ethanimidoate, ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate, kqcbswbqaxtilk-owbhpgmisa-n, z-ethyl n-mesitylsulfonyloxyacetimidate, z-ethyl n-mesitylsulfonyl oxyacetimidate | |
ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate |
Safety and Handling
EINECSNumber : 253-825-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only