missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Nonafluorobutyl Ether (mixture of isomers), TCI America™

Manufacturer:  TCI America E052825G

 View more versions of this product

Catalog No. E052825G



Specifications

Specifications

Ethyl Nonafluorobutyl Ether (mixture of isomers)
25g
C6H5F9O
DFUYAWQUODQGFF-UHFFFAOYSA-N
1-ethoxy-1,1,2,2,3,3,4,4,4-nonafluorobutane
206000
Clear Liquid at 20°C
Colorless
813458-04-7
Ethyl Perfluorobutyl Ether, Nonafluorobutyl Ethyl Ether, Perfluorobutyl Ethyl Ether
CCOC(C(C(C(F)(F)F)(F)F)(F)F)(F)F
264.091
264.09
≥98.0% (GC)

Chemical Identifiers

813458-04-7
264.091
Ethyl Perfluorobutyl Ether, Nonafluorobutyl Ethyl Ether, Perfluorobutyl Ethyl Ether
1-ethoxy-1,1,2,2,3,3,4,4,4-nonafluorobutane
C6H5F9O
DFUYAWQUODQGFF-UHFFFAOYSA-N
206000
CCOC(C(C(C(F)(F)F)(F)F)(F)F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents