missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Nonadecanoate 96.0+%, TCI America™
Supplier: TCI America N04595G
Specifications
Ethyl Nonadecanoate | |
White-Yellow | |
MFCD00026690 | |
Nonadecanoic Acid Ethyl Ester | |
CCCCCCCCCCCCCCCCCCC(=O)OCC | |
326.565 | |
326.57 | |
Crystals |
18281-04-4 | |
C21H42O2 | |
5 g | |
ICVYQLQYVCXJNE-UHFFFAOYSA-N | |
ethyl nonadecanoate | |
29008 | |
≥96.0% (GC) |
Chemical Identifiers
18281-04-4 | |
326.565 | |
ICVYQLQYVCXJNE-UHFFFAOYSA-N | |
29008 | |
CCCCCCCCCCCCCCCCCCC(=O)OCC |
C21H42O2 | |
MFCD00026690 | |
Nonadecanoic Acid Ethyl Ester | |
ethyl nonadecanoate |
Safety and Handling
TSCA : No