Please login to your online account to display your discounted pricing

Ethyl nitroacetate, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  185170010

 View more versions of this product

Catalog No. AC185170010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Formula Weight 133.1
Physical Form Liquid
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.199g/mL
Molecular Formula C4H7NO4
Refractive Index 1.4235 to 1.4255 (20°C, 589nm)
Assay Percent Range 98%
CAS 626-35-7
Linear Formula O2NCH2CO2C2H5
Infrared Spectrum Authentic
Chemical Name or Material Ethyl nitroacetate

Specifications

Chemical Name or Material Ethyl nitroacetate
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C4H7NO4
Linear Formula O2NCH2CO2C2H5
Formula Weight 133.1