Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl nipecotate, 97%

$51.38 - $121.43

Chemical Identifiers

Chemical Name or Material Ethyl nipecotate
CAS 5006-62-2
Molecular Formula C8H15NO2
Formula Weight 157.21
MDL Number MFCD00005991
Synonym Ethyl piperidine-3-carboxylate; Nipecotic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2415514

  SDS
alfa aesar™
B2415514
25g Each for $51.38

AAB2415522

  SDS
alfa aesar™
B2415522
100g Each for $121.43
Description & Specifications

Specifications

Formula Weight 157.21
Chemical Name or Material Ethyl nipecotate
Assay 97%
Molecular Formula C8H15NO2
MDL Number MFCD00005991
CAS 5006-62-2
Synonym Ethyl piperidine-3-carboxylate; Nipecotic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Flash Point 90°C (195°F)
Boiling Point 102° to 104°C (7mmHg)
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

2° to 8°C