missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Nicotinate 99.0+%, TCI America™
$59.73 - $449.59
Chemical Identifiers
CAS | 614-18-6 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.165 |
MDL Number | MFCD00006389 |
InChI Key | XBLVHTDFJBKJLG-UHFFFAOYSA-N |
Synonym | ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan |
PubChem CID | 69188 |
IUPAC Name | ethyl pyridine-3-carboxylate |
SMILES | CCOC(=O)C1=CN=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
N008525ML
|
TCI America
N008525ML |
25 mL |
Each for $59.73
|
|
|||||
N0085100ML
|
TCI America
N0085100ML |
100 mL |
Each for $149.32
|
|
|||||
N0085500ML
|
TCI America
N0085500ML |
500 mL |
Each for $449.59
|
|
|||||
Chemical Identifiers
614-18-6 | |
151.165 | |
XBLVHTDFJBKJLG-UHFFFAOYSA-N | |
69188 | |
CCOC(=O)C1=CN=CC=C1 |
C8H9NO2 | |
MFCD00006389 | |
ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan | |
ethyl pyridine-3-carboxylate |
Specifications
614-18-6 | |
224°C | |
MFCD00006389 | |
ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan | |
CCOC(=O)C1=CN=CC=C1 | |
151.165 | |
151.17 | |
Liquid |
Yellow | |
C8H9NO2 | |
25 mL | |
XBLVHTDFJBKJLG-UHFFFAOYSA-N | |
ethyl pyridine-3-carboxylate | |
69188 | |
≥99.0% (GC) | |
Ethyl Nicotinate |
Safety and Handling
EINECSNumber : (5)-6766
TSCA : Yes