missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-(tert-Butoxycarbonyl)-D-pyroglutamate, TCI America™

Manufacturer:  TCI America E09405G

 View more versions of this product

Catalog No. E09405G



Specifications

Specifications

Ethyl N-(tert-Butoxycarbonyl)-D-pyroglutamate
56°C
144978-35-8
MFCD09261329
YWWWGFSJHCFVOW-MRVPVSSYSA-N
1-O-tert-butyl 2-O-ethyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate
820885
Crystal-Powder at 20°C
White
5g
C12H19NO5
N-(tert-Butoxycarbonyl)-D-pyroglutamic Acid Ethyl Ester, Ethyl (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic Acid Ethyl Ester, Ethyl N-Boc-D-pyroglutamate, N-Boc-D-pyroglutamic Acid E
CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
257.286
257.29
≥95.0% (GC)

Chemical Identifiers

144978-35-8
257.286
YWWWGFSJHCFVOW-MRVPVSSYSA-N
820885
CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C
C12H19NO5
MFCD09261329
N-(tert-Butoxycarbonyl)-D-pyroglutamic Acid Ethyl Ester, Ethyl (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic Acid Ethyl Ester, Ethyl N-Boc-D-pyroglutamate, N-Boc-D-pyroglutamic Acid E
1-O-tert-butyl 2-O-ethyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents