missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-Phenylformimidate, TCI America™

Manufacturer:  TCI America E049525G

 View more versions of this product

Catalog No. E049525G



Specifications

Specifications

Ethyl N-Phenylformimidate
Yellow
6780-49-0
MFCD00043655
DRDBNKYFCOLNQO-UHFFFAOYSA-N
ethyl N-phenylmethanimidate
111236
Clear Liquid at 20°C
215°C
25g
C9H11NO
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide
CCOC=NC1=CC=CC=C1
149.193
149.19
≥98.0% (GC)

Chemical Identifiers

6780-49-0
149.193
DRDBNKYFCOLNQO-UHFFFAOYSA-N
111236
CCOC=NC1=CC=CC=C1
C9H11NO
MFCD00043655
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide
ethyl N-phenylmethanimidate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents