Learn More
Ethyl N-phenylformimidate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04378814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl N-phenylformimidate | |
Colorless | |
C9H11NO | |
25 g | |
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide | |
CCOC=NC1=CC=CC=C1 | |
149.19 | |
149.19 | |
Liquid |
6780-49-0 | |
101°C to 102°C (15 mmHg) | |
MFCD00043655 | |
Air and moisture sensitive | |
DRDBNKYFCOLNQO-UHFFFAOYSA-N | |
ethyl N-phenylmethanimidate | |
111236 | |
97% |
Chemical Identifiers
6780-49-0 | |
149.19 | |
DRDBNKYFCOLNQO-UHFFFAOYSA-N | |
111236 | |
CCOC=NC1=CC=CC=C1 |
C9H11NO | |
MFCD00043655 | |
ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide | |
ethyl N-phenylmethanimidate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 229-841-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only