missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-hydroxyacetimidate, 97%, ACROS Organics™

Manufacturer:  Acros Organics 197500500

 View more versions of this product

Catalog No. AC197500500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl N-hydroxyacetimidate
Authentic
1.4325 to 1.4345
Colorless
50g
10576-12-2
C4H9NO2
MFCD00002114
Solubility in water: may decompose.
CCOC(=NO)C
103.12
103.12
97%
77°C
Glass bottle
55.0°C to 58.0°C (6.0mmHg)
23.0°C to 25.0°C
97%
97%
CH3C(=NOH)OC2H5
1-ethoxyethanone oxime
QWKAVVNRCKPKNM-PLNGDYQASA-N
ethyl (1Z)-N-hydroxyethanimidate
6386647
Liquid After Melting

Chemical Identifiers

10576-12-2
103.12
QWKAVVNRCKPKNM-PLNGDYQASA-N
6386647
CCOC(=NO)C
C4H9NO2
MFCD00002114
1-ethoxyethanone oxime
ethyl (1Z)-N-hydroxyethanimidate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 234-165-7

TSCA : TSCA

SDS Documents